Toluene ≥99.5% ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

T1090-20LTMT T1090-100ML T1090-4LTGL T1090-500MLG T1090-1LTGL
700002-410EA 844.76 USD
700002-410 700002-406 700002-412 700002-414 700002-408
Toluene ≥99.5% ACS for synthesis
Toluene

Toluene, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: C₆H₅CH₃
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.865
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR