Silver sulfate 98, powder ACS for synthesis

Danger

Synonyms: Disilver sulfate, Silver(I) sulfate, Disilver sulphate

700002-086EA 1091.85 USD
700002-086 700002-088 700002-092 700002-090
Silver sulfate 98, powder ACS for synthesis
Silver sulphate

Silver Sulfate, Powder, Reagent, ACS is a silver compound used in silver plating and is minimally soluble in water. It is a black solid sulfate that is stable under common conditions.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085
Melting Pt: 655
Density: 5.450
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR