PolyGen™ Mops, Micronova
Supplier: Micronova
MZLM5-12IR PG9-12 MZLM5-12 PG9-12IR
68400-786CS 1610.44 USD
68400-786 68399-706 68400-758 43300-075
PolyGen™ Mops, Micronova
Mops
PolyGen™ mops are ideal for disinfecting in pharmaceutical and biotech environments.
  • Low-linting polyester looped strands with dual-action abrasive mesh
  • Herringbone weave ensures there are no raw edges, and limits particle generation
  • Mops are reuseable, launderable, and autoclavable
LoopMop™ mops are knitted polyester looped mops for applying disinfectant in pharmaceutical, aseptic processing, and medical device applications. The polyester material is absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants thatMore Product Information