Polywipe Extra Wipes, Contec®
Supplier: Contec
3022-99
68310-068CS 435.3 USD
68310-068 68310-070
Polywipe Extra Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polywipe Extra Wipes are unlaundered heavyweight polyester knit wipes with knife-cut edges that can be used in less critical applications.
  • Recommended for ISO Class 6 to 8 environments
  • Knitted 100% polyester fabric for low levels of particles and fibers
  • Soft edge is ideal for use on surfaces which are vulnerable to scratching
  • Abrasion and chemical resistant
  • Cost effective for equipment wipe down and general cleaning
  • Durable no run interlock knit construction keeps the fabric from unraveling
The durable, no-run interlock knit construction makes itMore Product Information