Nalgene® Imhoff Settling Cone and Cone Rack, Thermo Scientific
Supplier: Thermo Fisher Scientific
1000-0010
66186-001CS 548.55 USD
66186-001
Nalgene® Imhoff Settling Cone and Cone Rack, Thermo Scientific
Funnels Sedimentation Cones
For the determination of settleable matter in water and waste effluent
Polycarbonate cone is fully graduated externally from the vortex upward. Polypropylene screw closure allows user to read graduations to zero. Leakproof plug makes it possible to withdraw sediment. Easily cleaned with water and detergent. Autoclavable.
Durable, transparent acrylic cone rack (sold separately) holds three Imhoff cones securely. Wide base provides stability on lab bench.
Also used to measure sand content in well water.