Whirl-Pak® Sterilized Standard Bags, Nasco Sampling
Supplier: NASCO SAMPLING LLC
B00736 B00994 B01027 B00679 B01009 B01323 B00992 B01020 B01018 B00990 B01063
11216-056CS 2269.7 USD
11216-056 11216-090 11216-420 11216-012 11216-373 11216-422 66175-612 11216-409 11216-781 11216-796 11216-089 11216-424
Whirl-Pak® Sterilized Standard Bags, Nasco Sampling
Bags Sample Bags
Blended, virgin polyethylene film provides exceptional clarity, superior strength, and sterilized after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous perimeter seal: Maximum protection against
More Product Information