Nunc® Lab-Tek™ II Chamber Slide™ System, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
154453 154534 154526 154461
62407-290CS 1079.89 USD
62407-290 62407-296 62407-294 62407-292
Nunc® Lab-Tek™ II Chamber Slide™ System, Sterile, Thermo Scientific
Slides Chamber Slides
This system features removable, polystyrene chambers that are sealed to a 25x75mm non-fluorescent glass slide using biocompatible acrylic adhesive
Numbers along the border of the chamber and a Superfrost® writing area on the slide facilitate identification.
The sealant remains with the media chamber after detachment, leaving a hydrophobic border to define cell culture areas. A tabbed cover fits over the media chamber allowing gas exchange while minimizing evaporation. The slides are provided sterile with an RS wash to create a consistent growth surface for adherent cell lines.
Order Now
SPECIFICATIONS
Environmentally Preferable