Corning® BioCoat® Laminin Culture Dishes, Corning
Supplier: Corning
354452 354405
62405-708CS 242.7 USD
62405-708 62405-706
Corning® BioCoat® Laminin Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Laminin (LM), a major component of basement membranes, is a multifunctional glycoprotein that is used as a substrate to culture and maintain differentiated function of a wide variety of cells. Laminin has been shown in culture to stimulate neurite outgrowth, promote cell attachment, chemotaxis, cell differentiation, and neuronal survival.
  • Promotion of cell attachment and spreading
  • Induction of cell differentiation and neurite outgrowth
  • Increases proliferation of myoblasts
  • Studies of effects of laminin on cell behavior
  • Cell adhesion assays
  • Storage: 2 to 8 °
More Product Information