Humidity/Temperature Monitor, Sper Scientific
Supplier: Sper Scientific
800016C 800016
61220-512EA 209.27 USD
61220-512 490012-194
Humidity/Temperature Monitor, Sper Scientific
Hygrometers Digital Thermo-Hygrometers
Digital humidity temperature monitor with a large display.
  • Available with or without certification
Simultaneously monitors both temperature and humidity on an extra large 3-3/16 x 2-1/2" dual display which can be read from across a room. Monitor humidity and temperature in labs, chemical storage areas, clean rooms, stock rooms, warehouses, factories, greenhouses, and outdoors. Recall min/max temperature and min/max humidity readings over any time period or have it automatically reset itself every 24 hours. Stand or wall mount.
Certifications: N.I.S.More Product Information