VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
90010-36 90010-44
61161-350EA 52.21 USD
61161-350 62379-460
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Timers Digital Stopwatches
Perfect for lab and plant use, this LCD digital stopwatch keeps time up to 24 hours.
  • Fine time-keeping resolution
  • Water-resistant, shock-resistant ABS plastic housing
  • Different models are color-coded for water resistance
  • Traceable®
Timing resolution is 0.01 seconds for the first 30 minutes.
Housed in a water-resistant/shock-resistant or waterproof/shockproof ABS plastic case. Water-resistant model is black. Waterproof model is yellow in color.
Continues timing to 23 hours, 59 minutes, 59 seconds with a 1 second resolution. Accuracy is 0.01%.More Product Information