HiVeg™ Microbiological Culture Media, HiCrome™ MRSA Agar Base to Sabouraud Maltose Broth, HiMedia Laboratories
Supplier: HiMedia
MV1067-500G MV1086-500G MV1213-500G MV1453-500G MV1469-500G MV661-500G MV946-500G MV575-5KG MV575-10KG MV1245-500G MV064-500G MV1021-500G MV1024-500G MV1084-500G MV1139-500G MV133-100G MV539-500G MV995-500G MV1151-500G M1582-500G MV1023-500G MV1163-500G MV1164-500G MV1269-500G MV669-100G MV284-500G MV575-500G MV098-500G MV1003-500G MV1166-500G MV1274-500G MV1552-500G MV872-100G MV1552-2.5KG MV1552-5KG MV976-500G
95039-426EA 279.27 USD
95039-426 95039-434 95039-450 95039-484 95039-492 95039-540 95039-558 95040-540 95040-542 61000-592 95039-402 95039-410 95039-414 95039-432 95039-436 95039-474 95039-536 95039-566 61000-490 95021-294 95039-412 95039-442 95039-444 95039-456 95039-542 95039-530 61001-140 95039-404 95039-406 95039-446 95039-460 95039-502 95039-550 89228-108 89228-110 95039-564
HiVeg™ Microbiological Culture Media, HiCrome™ MRSA Agar Base to Sabouraud Maltose Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based media designed to work in conjunction with or replace standard animal-based protein sources
The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of media can meet the requirements of nearly any microbiology application.