Nalgene® Autoclaving Baskets, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
6917-0127 6917-0150 6917-0230
60915-390CS 232.41 USD
60915-390 60915-392 60915-394
Nalgene® Autoclaving Baskets, Polypropylene, Thermo Scientific
Baskets Safety Transporting Baskets
These baskets hold labware during autoclaving and washing
They allow complete venting and draining. Smooth surface will not scratch plastic or glass. White polypropylene will not rust. Nestable for storage.
Order Now
SPECIFICATIONS
Environmentally Preferable