Hydrion™, MicroFine pH Test Paper with Dispenser, Micro Essential Laboratory
Supplier: Micro Essential
MF-1602 MF-1603 MF-1604 MF-1606 MF-1607
60775-956CS 155.11 USD
60775-956 60775-958 60775-960 60775-964 60775-966
Hydrion™, MicroFine pH Test Paper with Dispenser, Micro Essential Laboratory
Test Paper pH Indicator Papers and Test Strips
This short-range Hydrion MicroFine pH Paper measures pH solutions in the acid pH range of 2.9-5.2 and offers clear bright single color matches at every 0.3 to 0.4 interval.
  • Provides precise, pinpoint pH measurement over a narrow pH range
  • Ultra-fast results with accuracy in intervals of 0.2 - 0.3 pH
  • Fit easily in to small tubes
These are ideally suited to testing minute amounts of solution, only requiring 0.0001mL for an accurate measurement.
Packaging: Packaged in a protective plastic dispenser with matching color chart contained in a polypropyleneMore Product Information