Synthware Solvent Still with Built-In Condenser, Kemtech America
Supplier: Kemtech America
S144125
60004-770EA 690.72 USD
60004-770
Synthware Solvent Still with Built-In Condenser, Kemtech America
Distillation Components Distillation Heads
Grease-free design.
  • Built in condenser
  • Standard 24/40 ground glass joint
  • Stopper and cap included
For excellent solvent and chemical resistance, the still is equipped with two 0-4mm high vacuum valves that have PTFE covers over the two Viton® O-rings on the shaft, and one integral PTFE O-ring on the tip, without exposing any material other than PTFE to any solvents. Includes 14/10 threaded joint and PTFE-lined septum for syringe access.