Synthware Manifold, Solvent Transfer
Supplier: Kemtech America
M164380
60004-598EA 218.47 USD
60004-598
Synthware Manifold, Solvent Transfer
Manifolds
Supplied with a high vacuum valve with a PTFE O-ring at the tip and PTFE wiper to protect Viton® O-ring on the shaft.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
For purifying and transfering solvents under inert conditions.