Synthware Distillation Head Adapter, Kemtech America
Supplier: Kemtech America
A542440
60000-642EA 415.08 USD
60000-642
Synthware Distillation Head Adapter, Kemtech America
Distillation Components Distillation Adapters
Synthware distillation head adapter is vacuum jacketed.
  • [ST] 10/18 top joint
  • Accommodates thermometer
  • Thermometer not included
The [ST] 10/18 top joint accommodates a 76mm immersion thermometer. Other [ST] joints are 24/40.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.