VWR® Red Rubber Stoppers, Flanged, Plasticoid
Supplier: VWR International
59586-300
59586-300PK 394.51 USD
59586-300
VWR® Red Rubber Stoppers, Flanged, Plasticoid
Stoppers Rubber Stoppers
Permit insertion of hypodermic needles.
This stopper fits five through 125 ml size bottles.
Order Now
SPECIFICATIONS
Environmentally Preferable