VWR® Rubber Stoppers
Supplier: VWR International
59586-157 59586-158 59586-159 59586-160 59586-161 59586-162 59586-163 59586-164 59586-167
59586-157CS 680.32 USD
59586-157 59586-158 59586-159 59586-160 59586-161 59586-162 59586-163 59586-164 59586-167
VWR® Rubber Stoppers
Stoppers Rubber Stoppers
Versatile rubber stoppers are four stoppers in one.
  • Easy conversion between stopper types
  • Versatile design
  • Soft and resilient
  • Natural rubber construction
  • Autoclavable
Three simple twist-off seals permit easy conversion from a solid to a one, two, or three-hole stopper for all your laboratory needs. Soft, resilient, natural, white rubber demonstrates exceptional aging characteristics and withstands repeated autoclaving.
Ordering information: Stoppers are available in sizes 3 to 10 or as an assortment of 33 stoppers. Sizes 3 and 4 only have two twist-More Product Information