Variable-Speed Electric Stirrers, Arrow
Supplier: Arrow Engineering
ARROW6000 ARROW1750 ARROW850 ARROW350-115
59025-124EA 1544.91 USD
59025-124 59025-126 59025-128 59025-130
Variable-Speed Electric Stirrers, Arrow
Stirrers Overhead Stirrers
These variable-speed, electronically controlled stirrers are built to handle viscosities ranging from water-like solutions to highly viscous fluids
Four models are available to handle a wide range of applications: Model 6000 is a low-cost, light-duty stirrer, ideal for stirring light to medium-light viscosity fluids. Model 1750 is a multi-purpose stirrer, designed to handle more viscous materials. Model 850 provides the speed and power to handle highly viscous materials. Its high-torque gear head maintains constant speed regardless of viscosity changes as mixing progresses. Slim design isMore Product Information