VWR® Spinstar® Magnetic Stir Bars
Supplier: VWR International
58949-026 58949-028 58949-032
58949-026CS 3264.41 USD
58949-026 58949-028 58949-032
VWR® Spinstar® Magnetic Stir Bars
Stirrers Magnetic Stirring Bars
Spinstar® magnetic stirring bars create a deep mixing vortex at relatively slow speeds
  • Star shape is designed to fit the inside diameter of most commonly-used beakers
  • Perfect for applications requiring slow, thorough mixing
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert.