Chex-All® II Instant-Seal Pouches, Propper Manufacturing
Supplier: Propper
02400800 02401000 02401200 02401800 02401400 02402800 02401500
58753-106CS 209.38 USD
58753-106 58753-139 58753-161 58753-194 58753-202 58753-220 76026-464
Chex-All® II Instant-Seal Pouches, Propper Manufacturing
Bags Sterilization Pouches
Instant self-seal sterilization pouches for use in steam autoclaves or ethylene oxide sterilizers provide a barrier to maintain sterility after the sterilization process.
  • Self-sealing sterilization Pouches for use in steam and ethylene oxide
  • Pre-printed indicator inside and outside the pouch
  • Features triple-sealed paper/plastic film to ensure the security of contents
  • Chemical indicator provides distinct color change from yellow to black in steam and also provides a distinct color change from red to green in EO
  • Made of 2.5 mm plastic and 73 to 74 gram paper
More Product Information