Staticide® Concentrate, ACL Staticide
Supplier: ACL Staticide
3000D 3000G 3000Q
58611-407EA 32723.95 USD
58611-407 58611-382 75780-008
Staticide® Concentrate, ACL Staticide
Surface Treatments Antistatic Treatments
Staticide® Concentrate is available to save freight, handling, and storage costs while offering greater flexibility in dilution.
Staticide® Concentrate can be diluted with tap waater, Isopropyl Alcohol, Carbon Tetrachloride, or Perchloroethylene to meet various processing needs.
Staticide® Concentrate should be mixed with water or other solvents in the following concentrate solvent ratios to obtain these formulations: General Purpose Staticide® 1:99, Heavy Duty Staticide® 1:39
Certifications: When properly applied, Staticide® complies with or exceeds all electrostatic decay criteriaMore Product Information