Lab Guard® Reclosable Biohazard Bags
Supplier: Minigrip
SBL2R69B SBL2X810B SBL2X1215B SBL2X69B5 SBL2X69B SBL2AP69B SBL2AP1215B SBL2X810B5 SBL2X69 SBL2X46B
56766-364CS 108.87 USD
56766-364 56766-372 56766-374 95059-670 56766-370 89403-312 89411-680 10124-476 10124-426 10124-430
Lab Guard® Reclosable Biohazard Bags
Bags Waste Disposal Bags
These specimen handling bags protect both workers and paperwork.
  • Re-closable zipper for secondary leakage containment
  • Additional marking areas identify contents needing to be kept frozen, refrigerated, or at room temperature
Many Lab Guard® bags now feature a double zipper for added closure security. Bags include document pouch.
They feature bright orange and black graphics to clearly identify bags as containing biohazardous materials. New destroyable biohazard symbol bags won't leak. They tear open easily at processing and can be disposed of inMore Product Information