Keystone Carbon Sampling Probe, Seedburo
Supplier: Seedburo Equipment
118-S
56724-056EA 3418.51 USD
56724-056
Keystone Carbon Sampling Probe, Seedburo
Samplers Solid Samplers
The Keystone Carbon Sampling probe features double tube construction, 13 openings along the side of the probe with partitions separating each compartment.
  • “T” welded handle
  • Made of chrome-plated steel