PIG® HazMat Mat Roll, New Pig
Supplier: New Pig
MAT309 MAT303 MAT320 MAT325
56620-086EA 223.95 USD
56620-086 56620-752 89205-908 89205-910
PIG® HazMat Mat Roll, New Pig
Spill Control Materials
Absorbs the widest range of acids, bases and unknown liquids - even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills. Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue.
  • Absorbs the widest
More Product Information