Nylon Hand Pump, New Pig
Supplier: New Pig
DRM462
56620-596EA 143.88 USD
56620-596
Nylon Hand Pump, New Pig
Pumps Drum/Transfer Pumps
Nylon hand pumps fits 15- through 55-gallon drums with 2" NPT bungs.
  • Moves 8 oz. of water-based liquid per stroke
  • Spout rotates upward to prevent messy drips between uses. Fits
  • Maximum recommended viscosity equals SAE 30 weight oil.