PIG® Heavy-Duty Poly Modular Spill Deck, New Pig
Supplier: New Pig
PAK466
56620-156EA 543.68 USD
56620-156
PIG® Heavy-Duty Poly Modular Spill Deck, New Pig
Drum Storage
Heavy-duty deck withstands extra-heavy loads; Excellent for storing or servicing heavy equipment that is prone to leaks
  • Low 4.25" height is easy to load or unload; pumps and funnels are easy to access
  • Catches leaks, drips and spills to help keep your storage areas clean and safe
  • Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
  • Textured grating adds traction and lifts out for easy sump access
Unique "flow-through" sump design lets liquid pass freely from one sump to another to help you comply withMore Product Information