Cease-Fire® Waste Receptacles, Justrite®
Supplier: Justrite
26630G 26612G 26430
56610-547EA 507.96 USD
56610-547 56610-553 56610-545
Cease-Fire® Waste Receptacles, Justrite®
Bins Waste Bins
For the safe accumulation of waste paper and other solvent-free combustibles. Contoured head directs smoke and gas from any fire inside the container back into the combustion area, cutting off the air supply and extinguishing flames in seconds. Provides round-the-clock protection against trash fires.
  • Specially contoured to direct smoke and gas from any fire inside the container, back into the combustion area, cutting off the air supply and extinguishing flames
  • Corrosion-free aluminum heads provide easy cleanup
  • Bodies are heavy-gauge steel with rust-
More Product Information