Cylinder Lockers, Justrite®
Supplier: Justrite
23002 23004 23006 23007 23008
56609-688EA 2432.35 USD
56609-688 56609-692 56609-696 56609-698 83007-892
Cylinder Lockers, Justrite®
Furniture Lockers
Safely secure gas cylinders, including liquefied petroleum gas (LPG) and oxygen cylinders, from theft and unauthorized access
  • Sloped roof to shed water and ice
  • Solid top to reinforce stability
  • Warning and full/empty cylinder labels
  • Extruded framework for extra strength
  • Padlock lug for security
  • Pre-drilled holes at each corner for anchoring
Horizontal lockers feature easy-to-use racks that position cylinders securely and can hold 9.1 kg (20 lbs.) or 15 kg (33 lbs.) cylinders. Vertical lockers feature a heavy-duty floor plate to keep cylinders clear ofMore Product Information