VWR® Neoprene Flask Underlay
Supplier: VWR International
56255-006
56255-006EA 31.74 USD
56255-006
VWR® Neoprene Flask Underlay
Supports Flask Supports
Moderate heat and oil resistance with concave recess on top to support round-bottom flasks and dishes.
  • Made of long-lasting green Neoprene rubber
Holds 250 and 500 mL flasks firmly in place.
Order Now
SPECIFICATIONS
Environmentally Preferable