Rockets® Reusable Earplugs, Moldex™
Supplier: Moldex/Metric
6400 6405 6420
56219-252CS 646.45 USD
56219-252 56219-254 56219-266
Rockets® Reusable Earplugs, Moldex™
Earplugs Reusable Earplugs
Reusable earplugs feature a brightly colored, easy grip handle for inserting plug into the ear.
  • An air bubble in the tip offers cushioned comfort all day long
  • Soft, flexible flanges provide a superior seal and fit most ear canal sizes
  • Available with or without connecting cord
  • Washable
Certifications: Meet ANSI 3.19-1974 standard.
Ordering information: Each pair comes with a Pocket-Pak® carrying case for hygienic storage of plugs not in use.
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health