Rhino Duo-Check Engine Coolant and Battery Test Refractometers, Reichert
Supplier: Reichert Technologies
137584L0
55611-002EA 458.83 USD
55611-002
Rhino Duo-Check Engine Coolant and Battery Test Refractometers, Reichert
Refractometers Handheld Refractometers
Handheld refractometers quickly and accurately determine freeze protection and percent concentration of engine coolants, battery charge level, and specific gravity
  • Automatic temperature compensation provides immediate, accurate direct readings of ethylene glycol, propylene glycol, and battery electrolyte with only a few drops of sample
  • Black, carbonate housing is dust, shock, and water resistant
  • Internal optics are sealed and cannot be contaminated. Six measurement scales
  • No batteries or power cord required
Antifreeze protection from –60 to 32 °F forMore Product Information