Convertors® TSM™ Sterility Maintenance Covers, Cardinal Health®
Supplier: Cardinal Health
T13812A T11622A T11630A T13015A T13610A T13220A T11015A T10610A T10812A
54110-586CS 306.86 USD
54110-586 54110-568 54110-570 54110-574 54110-580 54110-576 54110-564 54110-560 54110-562
Convertors® TSM™ Sterility Maintenance Covers, Cardinal Health®
Sterilization Wraps
Ideal for covering wrapped items after sterilization to ensure maximum protection from bacteria, moisture, and dust.
Made of polypropylene. Tear-resistant. Regular seal covers may be heat-sealed or taped for added security. Self-seal covers feature a pressure-sensitive closure that ensures the integrity of the seal. The Tower-Tear® snap-open feature allows opening of covers quickly and easily without scissors or other sharp instruments that could damage the contents.
Order Now
SPECIFICATIONS
Environmentally Preferable