Upchurch Scientific® Coned Port Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Supplier: Upchurch Scientific
VHP-320 VHP-320X VHP-325X F-300X F-330X F-130X F-142X F-192X F-195 F-120X F-192 F-193 U-321X U-400X U-320X F-132 LT-100X U-401X F-120 PK-120BLKX VHP-200 F-195X
89209-776EA 69.78 USD
89209-776 89209-778 89209-782 21511-078 14234-088 21511-054 21511-058 21511-666 21511-668 21511-861 21511-052 21511-664 21001-072 21511-304 21511-312 53500-822 53501-324 21511-083 21511-314 97055-078 97055-114 97055-140 21511-671
Upchurch Scientific® Coned Port Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Fittings and Connectors
RheFlex® Two-Piece Fingertight PEEK™ fittings provide inert, biocompatible connections for instrumentation
Each 1/16" fittings set contains a 10-32 threaded nut and a specially designed PEEK ferrule, and the 1/8" fittings sets a 5/16-24 PEEK nut and ferrule. Three lengths of the 1/16" nut are available; standard, short, and extra long. ReFlex Fingertight fittings are rated for use up to 7,000 psi (483 bar). Use ChromTRAC knobs (part number 6000-283) with RheFlex fittings for fingertight convenience and to color-code connections. Pressure rated up to 20,000 psi (1,380 bar) when wrenchMore Product Information