Sparging Relief Valve, Upchurch Scientific®
Supplier: Upchurch Scientific
P-753
53500-818EA 223.27 USD
53500-818
Sparging Relief Valve, Upchurch Scientific®
Valves Non Return Valves
The 3/16" ruby ball will quickly open and depressurize the reservoir before the mobile phase can rise up the tubing.