Falcon® Bulk Packaged Serological Pipettes, Corning
Supplier: Corning
357506 357508 357515 357530
53300-261CS 397.83 USD
53300-261 53300-365 53392-198 53300-501
Falcon® Bulk Packaged Serological Pipettes, Corning
Pipettes Serological Pipettes
The pipette graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.
  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0.1 EU/ml
  • Color-coded package and markings for ease of identification
Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduationsMore Product Information