VWR® Culture Tubes, Disposable, with Screw-Cap Finish, Borosilicate Glass
Supplier: VWR International
53283-685 53283-686 53283-800 53283-802 53283-804 53283-806
53283-685CS 413.81 USD
53283-685 53283-686 53283-800 53283-802 53283-804 53283-806
VWR® Culture Tubes, Disposable, with Screw-Cap Finish, Borosilicate Glass
Tubes Culture Tubes
Tubes have a marking spot and reference mark for orientation during use and a threaded neck to accept screw cap.
  • Borosilicate glass constuction
  • Marking spot and orientation reference mark on all tubes
Please note: The capacity given below indicates the brim capacity (overflow capacity).
Delivery information:
Caps are not supplied, sold separately.