3M™ White Super Polish Floor Pads 4100
Supplier: Janitorial Supplies
MMM08483
500039-351CT 16.66 USD
500039-351 500039-354
3M™ White Super Polish Floor Pads 4100
Wipes Cleaning Kits and Pads
For buffing very soft finishes or for polishing soft waxes on wood floors.
Removes scuffs and black heel marks. Enhances floor appearance.
Used with rotary machine speed of 175 to 600 rpm.
Order Now
SPECIFICATIONS
Environmentally Preferable