Accessories for Vacuum Ovens, Shel Lab
Supplier: Sheldon Manufacturing
3450706
490012-640EA 263.61 USD
490012-640
Accessories for Vacuum Ovens, Shel Lab
Ovens Vacuum Ovens
This gasket is a replacement for both a 9.2 x 9.2 and 9.3 x 9.3 gasket.
  • General and high temperature applications
  • Resistant to moderate or oxidizing chemicals, ozone, concentrated sodium hydroxide
  • Attacked by many solvents, oils, concentrated acides, dilute sodium hydroxide