Labnet Spectrafuge™ 16M Microcentrifuge, Labnet International
Supplier: Labnet International
C0160-SS C0160
490008-684EA 335.07 USD
490008-684 490014-072
Labnet Spectrafuge™ 16M Microcentrifuge, Labnet International
Centrifuges Microcentrifuges
The Spectrafuge 16M is the perfect, high speed, compact microcentrifuge. A brushless, maintenance free motor provides extremely fast acceleration and deceleration of the 18 place rotor and makes the Spectrafuge 16M the quietest microcentrifuge in its class.
  • Brushless motor, maintenance free
  • Exceptionally quiet
  • Extremely small footprint
  • Unique airflow patterns so samples stay cool
  • Cold room compatible
Speed and time are set with the control knobs on the front panel of the unit. A quick button allows for momentary runs. A continuous flow of ambient airMore Product Information