Diversified Biotech Wash-N-Dry™ Coverslip Rack
Supplier: Diversified Biotech
WSDR-1000
490007-150EA 65.03 USD
490007-150
Diversified Biotech Wash-N-Dry™ Coverslip Rack
Slide Holders
Holds up to ten 18, 22, and 25 mm slipcovers.
  • Fast and easy cleaning, surface-treating and drying
  • Includes a notch to secure coverslips with a rubber band or loop of monofilament
  • Fits inside a 100 mL beaker with space underneath for small stir bar when submerged
  • Chemically and thermally resistant
  • Autoclavable