Chemical-Resistant Labels, Diversified Biotech, Tough-Tags®
Supplier: Diversified Biotech
CLTT-1000 CLTT-2000 CLTT-3000 FLAG-1000 LTTI-1000 LTTI-2000 LTTM-1000 MISL-1000 RNBW-3000 RNBW-3100 RNBW-3200 SETT-1000 THER-1094 THER-1128 THER-1137 THER-1150 TTGP-1000 TTGP-1050 TTLB-1000 TTLC-1000 TTLG-1000 TTLO-1000 TTLR-1000 TTLW-1000 TTLW-2016 TTLW-2016-B TTLW-2016-G TTLW-2016-Y TTLW-KK-20 TTLY-1000 TTSB-1000 TTSC-1000 TTSG-1000 TTSR-1000 TTSW-1000 TTSW-2240 TT-SWALL TTSW-KK-20 TTSY-1000 TT-TNY TTSW-2240-B TTSW-2240-G TTSW-2240-O TTSW-2240-R TTSW-2240-Y TTGP-1050-B TTGP-1050-G TTGP-1050-O TTGP-1050-R TTGP-1050-Y MISL-1000-B MISL-1000-G MISL-1000-O MISL-1000-R MISL-1000-Y TTSH-1000 THER-3137
490006-698PK 82.41 USD
490006-698 490006-700 490006-702 490006-746 490006-848 490006-850 490006-852 490006-854 490006-930 490006-932 490006-934 490006-936 490006-976 490006-978 490006-980 490006-982 490007-054 490007-056 490007-058 490007-060 490007-062 490007-064 490007-066 490007-068 490007-070 490007-072 490007-074 490007-080 490007-082 490007-084 490007-092 490007-094 490007-096 490007-100 490007-102 490007-104 490007-106 490007-108 490007-110 490007-112 10141-662 10141-664 10141-666 10141-668 10141-670 10141-672 10141-674 10141-676 10141-678 10141-680 10141-682 10141-684 10141-686 10141-688 10141-690 10141-692 10141-718 10141-720 10141-722 10141-724 10141-726
Chemical-Resistant Labels, Diversified Biotech, Tough-Tags®
Labels
Tough-Tags® are chemically resistant, flexible, polyester labels that can withstand autoclaving, boiling water baths, freezing to –40 °C, or heating up to 121°C without degrading. The thermal transfer Tough-Tags can withstand up to 150 °C.
They adhere strongly to plastics and other materials. Tough-Tags® are thin and will not bind centrifuge rotors.