Cryogenic Printer Labels, Brady
Supplier: Brady Worldwide
116369 116380 116375 116393 116402 116387 116386 116372 116377 116371 142238 116367 116385 116381 116383 116389 116373 116418 116414 116379 116382 116384 116368 116376 116370 142207 142209 142208 142078 112497 114194 114202 114287 114288 114289 142075 114204 114200 114203 112499 142077 112495 112648 114201 121021 121029 121026 121018 121028 121022 121020 121019 121023 121024 121030 121025 121027 121017 121031 114532 114527 114529 THT-143-492-1.5-SC THT-152-492-2.5-SC THT-156-490-1.5-SC THT-158-490-1.5-SC THT-179-492-2.5-SC THT-250-492-1.5-SC THT-251-492-1.5-SC 114179 THT-59-492-2.5-SC 114222 116410 116408 116417 142076
89175-182RL 108.15 USD
89175-182 89175-184 89175-186 89175-196 89175-200 89175-202 89175-204 89175-206 89175-208 89175-210 89175-212 89175-214 89175-216 89175-222 89175-224 89175-226 89175-228 89175-240 89175-242 89175-244 89175-246 89175-248 89175-250 89175-252 89175-254 89175-262 89175-264 89175-266 89201-124 89098-186 89098-192 89125-012 89209-536 89209-542 89209-548 89209-550 89209-552 89209-574 89209-546 89209-538 89209-544 89209-572 89098-184 89098-188 89098-190 89209-560 89209-578 89098-182 89098-194 89209-540 89233-540 89233-544 89233-548 89233-552 89233-556 89233-560 89233-564 89233-568 89233-572 89233-576 89233-580 89233-584 89233-588 89233-592 89233-596 490006-278 490006-282 490006-288 490006-320 490006-324 490006-336 490006-344 490006-352 490006-360 490006-364 490006-366 490006-374 490006-382 490006-384 89175-221 89175-233 89175-239 89209-577
Cryogenic Printer Labels, Brady
Labels
Cryogenic labels for tubes, vials, and canes.
Chemical and stain resistant labels for slides and cassettes:
Temperatures from –80 to 121 °C (–112 to 250 °F)
B-449 Slide label with removability and write on capabilities
B-481 Stainerbondz™ extreme chemical resistant slide label
B-482 Tissue cassette label applied pre-process with the BSP®31 for extreme solvent/stain resistance
B-488 Slide label with moderate chemical
B-498 Tissue cassette label for re-identification of processed paraffin blocks.
General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more:
B-424More Product Information