GeneMate 0.65 ml Graduated Microcentrifuge Tubes for Boiling Applications, Scientific Specialties
Supplier: VWR International
490003-174 490003-176 490003-182 490003-184 490003-186 490003-188 490003-194 490004-456
490003-174BX 157.77 USD
490003-174 490003-176 490003-182 490003-184 490003-186 490003-188 490003-194 490004-456
GeneMate 0.65 ml Graduated Microcentrifuge Tubes for Boiling Applications, Scientific Specialties
Tubes Microcentrifuge Tubes
Tubes feature an improved easy open snap seal that will not pop open during boiling. An ergonomically designed cap makes these tubes easier to open and close than other tubes.
  • Positive flat cap seal design
  • Target syringe port
  • Autoclavable
  • Suitable for temperature range from –90 to 120 °C
  • Wide cap flange for easy opening
  • Rated to 20000 ×g
  • Free of detectable DNase, RNase, and human DNA
All tubes feature frosted writing surfaces on the cap and side for smear resistant labeling. Graduated tubes feature external graduations every 100 L. Molded fromMore Product Information