Bev-L-Edge® Micro Slides, Propper Manufacturing
Supplier: Propper
15300100 15400100
48300-364CS 294.11 USD
48300-364 48300-400
Bev-L-Edge® Micro Slides, Propper Manufacturing
Slides Microscope Slides
Bev-L-Edge® microscope slides are made of first quality water-white glass and are excellent for use with blood smears or in routine microscopy.
  • Top quality ready-to-use slides for use in routine microscopy
  • Features smoothly ground beveled edges
  • Single side frosted or double side Twin-Frosted
  • Available in thickness 0.9 mm - 1.0 mm
Bev-L-Edge® Microscope Slides are made of top quality, non-corrosive, non-fogging water-white glass. Slides are priced for single use, but durable enough for repeated applications. Bev-L-Edge® Microscope Slides are availableMore Product Information