SP Bel-Art Three-Way Fluoroplastic Stopcocks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F30888-0010
47750-034EA 166.01 USD
47750-034 47750-036 47750-038
SP Bel-Art Three-Way Fluoroplastic Stopcocks, Bel-Art Products, a part of SP
Stopcocks
The three-way configuration of these stopcocks permits connection of any two or all three ports.
  • PVDF construction
  • Chemically resistant
  • Autoclavable
Barbed tubulations accept rubber or plastic tubing. They are made of PVDF (polyvinylidene fluoride), a fluoroplastic resistant to most chemicals that withstands temperatures up to 204ºC (400ºF). Maximum pressure rating 34.3 KPa (5 psi). Steam autoclavable.