StatZorb® Wipers, Contec®
Supplier: Contec
SZ-99
47734-192CS 1773.78 USD
47734-192
StatZorb® Wipers, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
StatZorb® Wipes are made from polyester fabric with a conductive fiber knitted in a specially designed grid pattern that minimizes 'hot spots'. The antistatic properties are uniform on both sides of the wipe. The surface resistivity is 4.3×107. The static decay time is less than 1.5 seconds.
  • Recommended for ISO Class 3 to 5
  • Polyester fabric is extremely low in both particles and extractables
  • Knitted with a conductive fiber in a specially designed grid pattern that minimizes 'hot spots'
  • Antistatic fabric is abrasion and chemical resistant
The wipes areMore Product Information