SP Bel-Art Poxygrid® Pasteur Pipette Can Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18942-0000
47729-700EA 100.09 USD
47729-700
SP Bel-Art Poxygrid® Pasteur Pipette Can Rack, Bel-Art Products, a part of SP
Pipette Stands
Designed for the safe, convenient storage of Pasteur Pipettes or transfer pipettes.
  • Safe and convenient storage
  • Durable
  • Epoxy-coated steel wire construction
  • Chemical- and corrosion-resistant
  • Autoclavable
Individual compartments store up to four pipette sterilization cans, each with dimensions of 5.7 x 5.7 cm (2¼ x 2¼"). The durable rack sits flat on the benchtop and supports pipettes at an angle to allow for easy selection and use.
Epoxy-coated steel wire is chemical and corrosion resistant and steam autoclavable at 121°C (250°F). pipette cans not included.