VWR® Culture Tubes, Disposable, Borosilicate Glass
Supplier: VWR International
47729-566 47729-568 47729-570 47729-572 47729-574 47729-576 47729-578 47729-580 47729-583 47729-584 47729-586
47729-566CS 550.22 USD
47729-566 47729-568 47729-570 47729-572 47729-574 47729-576 47729-578 47729-580 47729-583 47729-584 47729-586
VWR® Culture Tubes, Disposable, Borosilicate Glass
Tubes Culture Tubes
VWR® Disposable culture tubes are resistant to chemical attack and weathering.
  • Chemically inert
  • High heat tolerance
  • Uniform dimensions
  • Convenient dispensing packages
Manufactured from Type I, Class B borosilicate glass, each tube is heat-treated at temperatures exceeding 500 °C (932 °F) immediately before packing, which eliminates contamination, increases strength, and reduces breakage.
Contents will not leach out and contaminate solutions. Borosilicate glass construction prevents pH change in laboratory tests.
Tubes exceed USP Type IMore Product Information