Baseline® Aneroid Sphygmomanometer Unit
Supplier: Fabrication Enterprises
12-2250
470357-422EA 32.5 USD
470357-422
Baseline® Aneroid Sphygmomanometer Unit
Blood Pressure Monitors
Baseline aneroid sphygmomanometer unit for blood pressure unit.
  • Circumference: 23 to 40 cm
  • Cuff material: Nylon cuff
  • Display type: Dial display
  • Number of tubes: 2-tubes
  • Size: Adult cuff
  • Style: Pocket aneroid
  • Target area: Arm
  • Usage: Reusable